Order Entry
Italy
ContactUsLinkComponent
IL-4, Mouse, Biotrak Assay, ELISA - 96 wells (Range: 11-500pg/ml. Sensitivity: 5pg/ml. Sample volume: 50ul. 1.5L wash buffer supplie d. Suitable for use with cell culture supernatant and serum samples) 1 * 96 Assays
  RPN2712
 :  Cytiva
IL-4, Mouse, Biotrak Assay, ELISA - 96 wells (Range: 11-500pg/ml. Sensitivity: 5pg/ml. Sample volume: 50ul. 1.5L wash buffer supplie d. Suitable for use with cell culture supernatant and serum samples) 1 * 96 Assays
  RPN2712
 :  Cytiva
Frequently Bought Together