Dichloroacetic acid (DCA) 99+%
5 ml
Dichloroacetic acid (DCA) 99+%
Codice catalogo ACRO11306-0050
Numero CAS: 79-43-6
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Dichloroacetic acid for synthesis 1 * 5 mL Codice catalogo8.03541.0005
Product 8.03541.0005 is similar to ACRO11306-0050 but manufacturer and specifications may vary.
Product Details & Documents
Synonyms: DCA Deblock, DCA
- CAS Number:79-43-6
- Formula:C₂H₂Cl₂O₂
- MDL Number:MFCD00004223
- Molecular Weight:128.94 g/mol
- Merck Index:13,03075
- Boiling Pt:194 °C (760 mmHg)
- Melting Pt:9…11 °C
- Density:1.56 g/cm³
- Flash Pt:>112 °C
- Storage Temperature:Ambient
- EINECS:201-207-0
- UN:1764
- ADR:8,II
Documents
ACRO11306
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4650 to 1.4670 (20°C, 589 nm)
- Color scale:≤100 APHA
- Titration with NaOH:99.0 to 101.0 %
Specifications
- Pk:5 ml



