1,2-Dibromoethane 99%
Glass bottle, 500 ml
Nome del fornitore: Thermo Fisher Scientific
1,2-Dibromoethane 99%
Codice catalogo ACRO112795000
Nome del fornitore: Thermo Fisher Scientific
Numero CAS: 106-93-4
Jump to:
Product Details & Documents
- CAS Number:106-93-4
- Formula:BrH₂CCH₂Br
- MDL Number:MFCD00000233
- Molecular Weight:187.86 g/mol
- Merck Index:13,03830
- Boiling Pt:131…132°C (760mmHg)
- Boiling Pt:131…132 °C (760 mmHg)
- Melting Pt:9…10°C
- Melting Pt:9…10 °C
- Density:2.173
- Flash Pt:>104°C
- Flash Pt:>104 °C
- Storage Temperature:Ambient
- EINECS:203-444-5
- UN:1605
- ADR:6.1,I
- ADR:6.1,
Specifications
Specification Test Results
- Appearance:clear colorless to light yellow liquid
- Infrared spectrum:Conforms
- GC:≥98.5%
- Refractive index:1.5375 to 1.5395 (20°C, 589nm)
Specifications
- Pack type:Glass bottle
- Pk:500 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



