Order Entry
Italy
ContactUsLinkComponent
Alcoholometer, EUROPA classII, measuring range:70-80:0.1%vol, without thermometer, length:360mm, reference temperature: 20C, suitab le for government verification. 1 * 1 items
Catalog # AMARH848124
Nome del fornitore:  Amarell
Alcoholometer, EUROPA classII, measuring range:70-80:0.1%vol, without thermometer, length:360mm, reference temperature: 20C, suitab le for government verification. 1 * 1 items
Catalog # AMARH848124
Nome del fornitore:  Amarell
Frequently Bought Together