Order Entry
Italy
Orders LinkContactUsLinkComponent
Cap strips, flat top
Cap strips, flat top
Strips of 12 flat caps, Clear, 25×8 strips
Catalog # 732-4897
Cap strips, flat top
Catalog # 732-4897
Cap strips, flat top
Cap strips, flat top
Strips of 12 flat caps, Clear, 25×8 strips
Price
108,00 €
Log in for availability
1 * 200 pezzi
Product Details & Documents
Made from polypropylene (PP).
  • Flat surface of each cap facilitates sample labelling
  • Large labelled end tabs for strip orientation and removal (A-H for 8 strip, 1 to 12 for 12 strip)
  • Certified free from DNase, RNase and Human DNA
Suitable for use with most of the 96-well PCR plates, as well as standard and low profile tube strips.
Colour: Natural
Specifications
  • Packed:
    25×8 strips
  • Colour:
    Clear
  • Description:
    Strips of 12 flat caps
  • Seq:
    03