Nitromethane 96% ACS
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:75-52-5
- Formula:CH₃NO₂
- MDL Number:MFCD00007400
- Molecular Weight:61.04 g/mol
- Merck Index:13,06644
- Boiling Pt:101.2 °C (1013 hPa)
- Melting Pt:–29 °C
- Density:1.12 g/cm³
- Flash Pt:35 °C
- Storage Temperature:Ambient
- EINECS:200-876-6
- UN:1261
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3805 to 1.3825 (20°C, 589 nm)
- Water:≤0.05 % (Coulometric)
- GC:≥95 %
- Color scale:≤10 APHA
Specifications
- Pack type:Glass bottle
- Pk:500 mL
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



