Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Calcein

Glass bottle, 5 g
Catalog # ACRO410630050
Irritant
Calcein
Catalog # ACRO410630050
CAS Number:  1461-15-0
Calcein
Calcein

Glass bottle, 5 g

CAS Number:  1461-15-0
Price
129,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193
  • CAS Number:1461-15-0
  • Formula:C₃₀H₂₆N₂O₁₃
  • MDL Number:MFCD00005049
  • Molecular Weight:622.53 g/mol
  • Melting Pt:200 °C
  • Storage Temperature:Ambient
  • EINECS:215-957-1
Specifications

Specification Test Results

  • Appearance:
    Yellow to orange-brown Powder
  • Infrared spectrum:
    Conforms
  • UV spectrum:
    Complies
  • Suitability:
    (for Ca determination) Passes test

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.