Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Watch glasses

Watch glass, PTFE, 75 mm
  216-1155
 :   Brand
Watch glasses
  216-1155
 :  Brand
Watch glasses
Watch glasses

Watch glass, PTFE, 75 mm

 
21,70 €
   
1 * 1 pezzi
 

PTFE, white.

  • Very good chemical resistance
  • Temperature resistant from –200 to +280 °C
  • Exceptionally smooth surfaces to prevent contamination

Ideal for covering PTFE beakers when decomposing and evaporating, can also be used as evaporating dishes. Extremely robust.

 
  • Ø:
    75 mm
  • Description:
    Watch glass, PTFE
  • Seq:
    06