Order Entry
Italy
Orders LinkContactUsLinkComponent
Stoppers, rubber

Stoppers, rubber

20,0 mm, 14,0 mm, 18,0 mm
Catalog # BRND144386
Supplier:   Brand
Stoppers, rubber
Catalog # BRND144386
Supplier:  Brand
Stoppers, rubber
Stoppers, rubber

20,0 mm, 14,0 mm, 18,0 mm

Price
25,90 €
Log in for availability
1 * 25 pezzi
Product Details & Documents

Natural rubber, red.

Specifications
  • Ø bottom:
    14,0 mm
  • Ø top:
    18,0 mm
  • Height:
    20,0 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.