1,2-Dibromobenzene 98%
Glass bottle, 25 mL
Supplier: Thermo Fisher Scientific
1,2-Dibromobenzene 98%
Catalog # ACRO348930250
Supplier: Thermo Fisher Scientific
CAS Number: 583-53-9
Jump to:
Product Details & Documents
- CAS Number:583-53-9
- Formula:C₆H₄Br₂
- MDL Number:MFCD00000057
- Molecular Weight:235.91 g/mol
- Boiling Pt:224 °C (760 mmHg)
- Melting Pt:4…6 °C
- Density:1.956 g/cm³
- Flash Pt:91 °C
- Storage Temperature:Ambient
- EINECS:209-507-3
- UN:3082
- ADR:9,III
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow to light pink Liquid
- Infrared spectrum:Conforms
- Refractive index:1.6100 to 1.6120 (20°C, 589 nm)
- GC:≥97.5 %
Specifications
- Pack type:Glass bottle
- Pk:25 mL



