Order Entry
Italy
Orders LinkContactUsLinkComponent
Polysorbate 40

Polysorbate 40

Glass bottle, 1 L
Catalog # ACRO334140010
Polysorbate 40
Catalog # ACRO334140010
CAS Number:  9005-66-7
Polysorbate 40
Polysorbate 40

Glass bottle, 1 L

CAS Number:  9005-66-7
Price
78,00 €
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
  • CAS Number:9005-66-7
  • Formula:C₆₂H₁₂₂O₂₆
  • MDL Number:MFCD00165345
  • Boiling Pt:? 100 °C (1013 hPa)
  • Melting Pt:~ 0,1 °C
  • Density:1.05 g/cm³
  • Flash Pt:>200 °C
  • Storage Temperature:Ambient

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear to hazy light yellow to dark yellow Viscous liquid or paste
  • Infrared spectrum:
    Conforms
  • Acid value:
    ≤3 mg KOH/g
  • Hydroxyl value:
    89 to 105 mg KOH/g
  • Saponification value:
    41 to 55 mg KOH/g

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications