Order Entry
Italy
Orders LinkContactUsLinkComponent
Double bossheads

Double bossheads

Double bosshead, thumb screw M8, Aluminium, 16,5 mm
Catalog # BOCH5350
Supplier:   BOCHEM
Double bossheads
Catalog # BOCH5350
Supplier:  BOCHEM
Double bossheads
Double bossheads

Double bosshead, thumb screw M8, Aluminium, 16,5 mm

Price
17,10 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Double bossheads, either made of stainless steel or aluminium.

  • Angle: 180 + 90°
  • Different screw types available
Specifications
  • Description:
    Double bosshead, thumb screw M8
  • Material:
    Aluminium
  • For rod Ø:
    16,5 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.