Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Retort rods, with M10 thread

12 mm, 600 mm, M10
Codice catalogo BOCH5111
Nome del fornitore:   BOCHEM
Retort rods, with M10 thread
Codice catalogo BOCH5111
Nome del fornitore:  BOCHEM
Retort rods, with M10 thread
Retort rods, with M10 thread

12 mm, 600 mm, M10

Price
25,10 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Stainless steel 18/10.

  • Rods for stand bases
  • M10 thread
Specifications
  • Ø:
    12 mm
  • Thread:
    M10
  • Length:
    600 mm
  • Seq:
    02