Order Entry
Italy
Orders LinkContactUsLinkComponent
Bromophenol red, pure

Bromophenol red, pure

Glass bottle for solids, 25 g
Catalog # ACRO304550250
Bromophenol red, pure
Catalog # ACRO304550250
CAS Number:  2800-80-8
Bromophenol red, pure
Bromophenol red, pure

Glass bottle for solids, 25 g

CAS Number:  2800-80-8
Price
236,00 €
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
  • CAS Number:2800-80-8
  • Formula:C₁₉H₁₂Br₂O₅S
  • MDL Number:MFCD00023014
  • Molecular Weight:512.18 g/mol
  • Melting Pt:230 °C
  • EINECS:220-538-1

Documents

Specifications

Specification Test Results

  • Appearance:
    Red-brown to brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 % (110°C)
  • Solubility:
    (1 % in ethanol) Clear
  • Visual transition interval:
    from pH 5.2 (yellow-red) to pH 7.0 (purple)

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.