Order Entry
Italy
Orders LinkContactUsLinkComponent
Counting chambers, Bürker
Counting chambers, Bürker
Counting chamber with clips, bright-lined, Bürker, 0,1 mm
Catalog # 612-5729
Supplier:   HIRSCHMANN
Counting chambers, Bürker
Catalog # 612-5729
Supplier:  HIRSCHMANN
Counting chambers, Bürker
Counting chambers, Bürker
Counting chamber with clips, bright-lined, Bürker, 0,1 mm
Price
169,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
These counting chambers with double net ruling are suitable for in vitro diagnostic applications.
Certifications: In vitro Diagnostic Devices Directive (98/79/EC)
Ordering information: Includes two cover glasses.
Specifications
  • Depth of chamber:
    0,1 mm
  • Type:
    Bürker
  • Description:
    Counting chamber with clips, bright-lined