Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Bulb pipettes, one mark, class B

4 ml, 0,023 ± ml, 400 mm, √
Catalog # 612-3464
Supplier:   HIRSCHMANN
Bulb pipettes, one mark, class B
Catalog # 612-3464
Supplier:  HIRSCHMANN
Bulb pipettes, one mark, class B
Bulb pipettes, one mark, class B

4 ml, 0,023 ± ml, 400 mm, √

Price
7,60 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

AR-Glas®

  • Calibrated to deliver (TD, Ex)
  • With silver diffusion ink durable brown graduations

DIN 12690

 : DIN EN ISO 648; DE-M marking

Specifications
  • Capacity:
    4 ml
  • Length:
    400 mm
  • Suppl. to DIN:
  • Tolerance:
    0,023 ± ml