Double spatulas, micro
130 mm, 28 mm, 5 mm
HAMMHWN029-13
Double-ended micro spatulas made from stainless steel WIRONIT steel.
- Length:130 mm
- Seq:02
- Blade width:5 mm
- Blade length:28 mm
Double-ended micro spatulas made from stainless steel WIRONIT steel.