Order Entry
Italy
Orders LinkContactUsLinkComponent
Forceps, teethed

Forceps, teethed

145 mm
Catalog # 232-2190
Supplier:   HAMMACHER KARL
Forceps, teethed
Catalog # 232-2190
Supplier:  HAMMACHER KARL
Forceps, teethed
Forceps, teethed

145 mm

Price
81,50 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

WIRONIT steel (18/12 stainless steel with 18% chrome and 10 - 12 % nickel)

  • Teethed, straight
  • Resistant to corrosion
  • Anti-magnetic
Specifications
  • Length:
    145 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.