Order Entry
Italy
Orders LinkContactUsLinkComponent
Microscopic forceps

Microscopic forceps

115 mm
Catalog # 232-0066
Supplier:   HAMMACHER KARL
Microscopic forceps
Catalog # 232-0066
Supplier:  HAMMACHER KARL
Microscopic forceps
Microscopic forceps

115 mm

Price
67,40 €
Log in for availability
1 * 1 pezzi
Product Details & Documents

Wironit steel (18/12 stainless steel with 18% chrome and 10 - 12% nickel).

  • Delicate
  • Resistant to corrosion
  • Anti-magnetic
Specifications
  • Length:
    115 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.