Order Entry
Italy
Orders LinkContactUsLinkComponent
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 160 mm
Catalog # HAMML-HWC053-16
Supplier:   HAMMACHER KARL
Thumb forceps, standard
Catalog # HAMML-HWC053-16
Supplier:  HAMMACHER KARL
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 160 mm
Price
63,90 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Standard forceps made from Wironit steel.
Specifications
  • Type:
    Straight, serrated, blunt
  • Length:
    160 mm
  • Seq:
    04