Order Entry
Italy
Orders LinkContactUsLinkComponent
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 130 mm
Catalog # HAMML-HWC051-13
Supplier:   HAMMACHER KARL
Thumb forceps, standard
Catalog # HAMML-HWC051-13
Supplier:  HAMMACHER KARL
Thumb forceps, standard
Thumb forceps, standard
Straight, serrated, blunt, 130 mm
Price
57,60 €
Log in for availability
1 * 1 pezzi
Product Details & Documents
Standard forceps made from Wironit steel.
Specifications
  • Type:
    Straight, serrated, blunt
  • Length:
    130 mm
  • Seq:
    02