Knife spatulas, flexible
180 mm, 80 mm
Catalog # HAMMHSN220-08
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Straight knife spatulas with wooden handle and stainless steel blade.
Specifications
- Length:180 mm
- Seq:08
- Blade length:80 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



