Grasping and dressing forceps, Gross-Maier
Gross-Maier, with catch, Straight, 250 mm
Catalog # HAMMHSE774-25
Supplier: HAMMACHER KARL
Jump to:
Product Details & Documents
Specifications
- Type:Straight
- Description:Gross-Maier, with catch
- Length:250 mm
- Seq:02