Dressing forceps, delicate
Delicate model, Straight, blunt, 145 mm
Codice catalogo HAMMHSC074-14
Nome del fornitore: HAMMACHER KARL
Jump to:
Product Details & Documents
Delicate dressing forceps made from stainless steel.
Specifications
- Type:Straight, blunt
- Description:Delicate model
- Length:145 mm
- Seq:03
- Cat. No.:HAMMHSC074-14