Order Entry
Italy
Orders LinkContactUsLinkComponent
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes

Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes

0,5 ml, Amber, With attached lid, Graduated, conical bottom, non sterile, 500 tubes (1 bag×500 tubes)
Catalog # 211-2145
   Low Manufacturing Impact
Supplier:   EPPENDORF
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Catalog # 211-2145
Supplier:  EPPENDORF
   Low Manufacturing Impact
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes
Eppendorf® Safe-Lock Tubes, 0,5 ml - Microtubes

0,5 ml, Amber, With attached lid, Graduated, conical bottom, non sterile, 500 tubes (1 bag×500 tubes)

Price
75,00 €
Log in for availability
1 * 500 pezzi
Product Details & Documents

Eppendorf® Safe-Lock tubes 0,5 ml - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

  • Hinged Safe-Lock lid prevents unintended opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss in the event of tube breakage, and provides enhanced safety when handling hazardous samples
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures
  • Precise lid sealing minimises evaporation rates during long-term storage
  • Large frosted lid and side surface facilitate easy labeling
  • Ensured functionality in a temperature range from −86 to +100 °C
  • Autoclavable when open (121 °C, 20 min.) for effective sterilisation
  • Available in lot-certified PCR clean, Eppendorf Forensic DNA grade, and Biopur® quality, ensuring tested purity

Eppendorf® Safe-Lock 0,5 ml microtubes epitomise advanced sample storage, featuring a patented Safe-Lock design for unparalleled security. Tailored for PCR applications, these microtubes provide a space-efficient 0,5 ml capacity, ideal for labs with limited storage. Their compatibility with various centrifuge models ensures seamless integration into existing workflows. Robust construction and user-friendly attributes, such as clear graduations, further enhance reliability and ease of handling. Elevate laboratory practices with Eppendorf® Safe-Lock tubes, which set a new standard for sample integrity and operational efficiency.

Manufactured without the use of slip agents, plasticisers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Cap type: Safe-Lock
Material: PP
Volume: 0,5 ml

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.

Specifications
  • Packed:
    500 tubes (1 bag×500 tubes)
  • Type:
    With attached lid
  • Version:
    Graduated, conical bottom, non sterile
  • Capacity:
    0,5 ml
  • Colour:
    Amber
  • Seq:
    09
  • Cat. No.:
    211-2145
  • Environmental certification:
    Renewable Energy Certificate, ISO 14001
  • Temperature range:
    −86…+100 °C
  • Autoclavable:
    When open 121 °C, 20 mins
  • Environmentally Preferable:
    Low Manufacturing Impact
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.