Vial racks, WHEATON®
17,1 mm, 90, 320×170×30 mm
Catalog # 548-2158
Supplier: DWK Life Sciences
Jump to:
Product Details & Documents
These PP racks accommodate 48, 50, 90 or 96 vials of different sizes.
- Autoclavable and stackable
- Good chemical resistance
- Includes position identifiers
Specifications
- Hole Ø:17,1 mm
- No. of Holes:90
- Seq:03
- W×D×H:320×170×30 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



