Order Entry
Italy
Orders LinkContactUsLinkComponent
These various racks are used with current and legacy Gilson liquid handlers, ASPEC® systems, and other instruments. The racks come i n a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates. 1 * 1 items
These various racks are used with current and legacy Gilson liquid handlers, ASPEC® systems, and other instruments. The racks come i n a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates. 1 * 1 items
Catalog # GILS2504611
New Product
Supplier:   GILSON
These various racks are used with current and legacy Gilson liquid handlers, ASPEC® systems, and other instruments. The racks come i n a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates. 1 * 1 items
Catalog # GILS2504611
Supplier:  GILSON
New Product
These various racks are used with current and legacy Gilson liquid handlers, ASPEC® systems, and other instruments. The racks come i n a variety of configurations to accommodate many different vials, tubes, microtubes, and multi-well microplates. 1 * 1 items
Price
271,00 €
Log in for availability
1 * 1 pezzi
Product Details & Documents