Order Entry
Italy
Orders LinkContactUsLinkComponent
Test tubes without rim

Test tubes without rim

160 mm, 16 mm, 1 mm
Codice catalogo HECH42775051
New Product
Nome del fornitore:   GLASWARENFABRIK KARL HECHT
Test tubes without rim
Codice catalogo HECH42775051
Nome del fornitore:  GLASWARENFABRIK KARL HECHT
New Product
Test tubes without rim
Test tubes without rim

160 mm, 16 mm, 1 mm

Price
Inquire For Price
Log in for availability
1 * 250 pezzi
Product Details & Documents

AR-Glas® (soda-lime glass).

  • Heavy wall (212-1700 suitable as Durham-Tube)
  • Good chemical and physical properties
  • Can also be used as disposable test tubes

Documents

Specifications
  • Ext.Ø:
    16 mm
  • Wall thickness:
    1 mm
  • Length:
    160 mm
  • Seq:
    17