Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Strata C18-E SPE Tubes

C18-E, PP, 70 Å, 55 µm, 12 ml, 2000 mg
Catalog # PHEC8B-S001-KDG
New Product
Supplier:   PHENOMENEX
Strata C18-E SPE Tubes
Catalog # PHEC8B-S001-KDG
Supplier:  PHENOMENEX
New Product
Strata C18-E SPE Tubes
Strata C18-E SPE Tubes

C18-E, PP, 70 Å, 55 µm, 12 ml, 2000 mg

Price
224,00 €
Log in for availability
1 * 20 pezzi
Product Details & Documents
Specifications
  • Particle Size:
    55 µm
  • Pore size:
    70 Å
  • Capacity:
    12 ml
  • Bed weight:
    2000 mg
  • Column hardware:
    PP
  • Phase:
    C18-E