Order Entry
Italy
Orders LinkContactUsLinkComponent
Strata C18-E SPE Tubes

Strata C18-E SPE Tubes

C18-E, PP, 70 Å, 55 µm, 1 ml, 50 mg
Catalog # PHEC8B-S001-DAM
New Product
Supplier:   PHENOMENEX
Strata C18-E SPE Tubes
Catalog # PHEC8B-S001-DAM
Supplier:  PHENOMENEX
New Product
Strata C18-E SPE Tubes
Strata C18-E SPE Tubes

C18-E, PP, 70 Å, 55 µm, 1 ml, 50 mg

Price
1 076,00 €
Log in for availability
1 * 500 pezzi
Product Details & Documents
Specifications
  • Particle Size:
    55 µm
  • Pore size:
    70 Å
  • Capacity:
    1 ml
  • Bed weight:
    50 mg
  • Column hardware:
    PP
  • Phase:
    C18-E
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.