Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Methoxy-1-Naphthaldehyde 98%

2-Methoxy-1-Naphthaldehyde 98%

Bottle
Catalog # APOSOR10105172.5KG
New Product
Supplier:   Apollo Scientific
2-Methoxy-1-Naphthaldehyde 98%
Catalog # APOSOR10105172.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  5392-12-1
2-Methoxy-1-Naphthaldehyde 98%

Bottle

CAS Number:  5392-12-1
Price
2 540,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents

Usage: Organic building block/reagent

  • CAS Number:5392-12-1
  • Formula:C₁₂H₁₀O₂
  • MDL Number:MFCD00004004
  • Molecular Weight:186,21 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications