Order Entry
Italy
Orders LinkContactUsLinkComponent
Phenol Red indicator

Phenol Red indicator

Bottle
Catalog # APOSOR1014625-10KG
Irritant
New Product
Supplier:   Apollo Scientific
Phenol Red indicator
Catalog # APOSOR1014625-10KG
Supplier:  Apollo Scientific
New Product
CAS Number:  143-74-8
Phenol Red indicator

Bottle

CAS Number:  143-74-8
Price
1 940,00 €
Log in for availability
1 * 10 kg
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide
  • CAS Number:143-74-8
  • Formula:C₁₉H₁₄O₅S
  • MDL Number:MFCD00003552
  • Molecular Weight:354,38 g/mol
  • EINECS:205-609-7
Specifications

Specifications

  • Pack type:
    Bottle

Specifications