Order Entry
Italy
Orders LinkContactUsLinkComponent
 

1,1-Dimethoxyacetone

Bottle
Catalog # APOSOR40044-10KG
Flammable
New Product
Supplier:   Apollo Scientific
1,1-Dimethoxyacetone
Catalog # APOSOR40044-10KG
Supplier:  Apollo Scientific
New Product
CAS Number:  6342-56-9
1,1-Dimethoxyacetone

Bottle

CAS Number:  6342-56-9
Price
1 870,00 €
Log in for availability
1 * 10 kg
Product Details & Documents
Synonyms:  Pyruvaldehyde 1,1-dimethyl acetal, 1,1-Dimethoxypropan-2-one, Pyruvic aldehyde dimethyl acetal
  • CAS Number:6342-56-9
  • Formula:C₅H₁₀O₃
  • MDL Number:MFCD00008758
  • Molecular Weight:118.13 g/mol
  • Boiling Pt:133…135 °C
  • Melting Pt:–57 °C
  • Density:1 g/cm³ (25 °C)
  • Flash Pt:37 °C
  • Storage Temperature:Ambient
  • UN:1224
  • ADR:3,III
Specifications

Specifications

  • Pack type:
    Bottle

Specifications