Inserisci ordine
Italy
Orders LinkContactUsLinkComponent
 

2-Naphthaldehyde

Bottle
Catalog # APOSOR350292-1G
Irritant
New Product
Supplier:   Apollo Scientific
2-Naphthaldehyde
Catalog # APOSOR350292-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  66-99-9
2-Naphthaldehyde

Bottle

CAS Number:  66-99-9
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  β-Naphthaldehyde
  • CAS Number:66-99-9
  • Formula:C₁₁H₈O
  • MDL Number:MFCD00004094
  • Molecular Weight:156.18 g/mol
  • Boiling Pt:160…19mm °C
  • Melting Pt:58…61 °C
  • Flash Pt:>100 °C
  • Storage Temperature:Congelamento
  • EINECS:200-640-2
  • UN:3335
  • ADR:9,III
Specifications

Specifications

  • Tipo di conf.:
    Bottle

Specifications