1-Hydroxy-2-naphthaldehyde
Bottle
Catalog # APOSOR18562-250MG
New Product
Supplier: Apollo Scientific
1-Hydroxy-2-naphthaldehyde
Catalog # APOSOR18562-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 574-96-9
Jump to:
Product Details & Documents
- CAS Number:574-96-9
- Formula:C₁₁H₈O₂
- MDL Number:MFCD00092325
- Molecular Weight:172.1800 g/mol
- Melting Pt:56 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



