Order Entry
Italy
Orders LinkContactUsLinkComponent
1-Hydroxy-2-naphthaldehyde

1-Hydroxy-2-naphthaldehyde

Bottle
Catalog # APOSOR18562-100G
Irritant
New Product
Supplier:   Apollo Scientific
1-Hydroxy-2-naphthaldehyde
Catalog # APOSOR18562-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  574-96-9
1-Hydroxy-2-naphthaldehyde

Bottle

CAS Number:  574-96-9
Price
2 050,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:574-96-9
  • Formula:C₁₁H₈O₂
  • MDL Number:MFCD00092325
  • Molecular Weight:172.1800 g/mol
  • Melting Pt:56 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.