Decoquinate 98%
Bottle
Catalog # APOSOR1030278-500G
New Product
Supplier: Apollo Scientific
Decoquinate 98%
Catalog # APOSOR1030278-500G
Supplier: Apollo Scientific
New Product
CAS Number: 18507-89-6
Jump to:
Product Details & Documents
Usage: Organic building block/reagent
Synonyms: Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
- CAS Number:18507-89-6
- Formula:C₂₄H₃₅NO₅
- Molecular Weight:417,54 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



