Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Decoquinate 98%

Bottle
Catalog # APOSOR1030278-1G
New Product
Supplier:   Apollo Scientific
Decoquinate 98%
Catalog # APOSOR1030278-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  18507-89-6
Decoquinate 98%

Bottle

CAS Number:  18507-89-6
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  Ethyl 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylate, Ethyl 6-(decyloxy)-7-ethoxy-4-hydroxyquinoline-3-carboxylate, 6-Decyloxy-7-ethoxy-4-hydroxyquinoline-3-carboxylic Acid Ethyl Ester
  • CAS Number:18507-89-6
  • Formula:C₂₄H₃₅NO₅
  • Molecular Weight:417,54 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications