Order Entry
Italy
Orders LinkContactUsLinkComponent
Bromochlorophenolblue 95% (dye content)

Bromochlorophenolblue 95% (dye content)

Bottle
Catalog # APOSOR1028099-250M
New Product
Supplier:   Apollo Scientific
Bromochlorophenolblue 95% (dye content)
Catalog # APOSOR1028099-250M
Supplier:  Apollo Scientific
New Product
CAS Number:  2553-71-1
Bromochlorophenolblue 95% (dye content)

Bottle

CAS Number:  2553-71-1
Price
36,20 €
Log in for availability
1 * 250 mg
Product Details & Documents

Usage: Organic building block/reagent

Synonyms:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromo-6-chlorophenol] S,S-dioxide
  • CAS Number:2553-71-1
  • Formula:C₁₉H₁₀Br₂Cl₂O₅S
  • Molecular Weight:581,06 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.