1-Nitro-2-naphthaldehyde 99%
Bottle
Codice catalogo APOSOR92158-25G
New Product
Nome del fornitore: Apollo Scientific
1-Nitro-2-naphthaldehyde 99%
Codice catalogo APOSOR92158-25G
Nome del fornitore: Apollo Scientific
New Product
Numero CAS: 101327-84-8
Jump to:
Product Details & Documents
Usage: Organic building block/ reagent.
Synonyms: 1-Nitro-2-naphthaldehyde
- CAS Number:101327-84-8
- Formula:C₁₁H₇NO₃
- MDL Number:MFCD03427164
- Molecular Weight:201,18 g/mol
Specifications
Specifications
- Pack type:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



