Order Entry
Italy
Orders LinkContactUsLinkComponent
α-Vitamin E ≥98%

α-Vitamin E ≥98%

Bottle
Catalog # APOSBUP14597-10G
New Product
Supplier:   Apollo Scientific
α-Vitamin E ≥98%
Catalog # APOSBUP14597-10G
Supplier:  Apollo Scientific
New Product
CAS Number:  59-02-9
α-Vitamin E ≥98%

Bottle

CAS Number:  59-02-9
Price
734,00 €
Log in for availability
1 * 10 g
Product Details & Documents

Usage: Biochemical/ life science applications.

Synonyms:  (+)-α-Tocopherol, Vitamin E Alcohol, (R)-2,5,7,8-Tetramethyl-2-((4R,8R)-4,8,12-trimethyltridecyl)chroman-6-ol, α-Vitamin E, 3,4-Dihydro-2,5,7,8-tetramethyl-2-(4,8,12-trimethyltridecyl)-2H-benzopyran-6-ol, Vitamin E, D(+)-α-Tocopherol
  • CAS Number:59-02-9
  • Formula:C₂₉H₅₀O₂
  • Molecular Weight:430,71 g/mol
Specifications

Specifications

  • Pack type:
    Bottle

Specifications