Order Entry
Italy
Orders LinkContactUsLinkComponent
4-Methoxy-1-naphthaldehyde 95%
Bottle
  APOSOR480638-5G
New Product
4-Methoxy-1-naphthaldehyde 95%
  APOSOR480638-5G
New Product
 :  15971-29-6
4-Methoxy-1-naphthaldehyde 95%
Bottle
 :  15971-29-6
21,80 €
1 * 5 g
 :  4-Methoxynaphthalene-1-carbaldehyde
  •  :15971-29-6
  •  :C₁₂H₁₀O₂
  •  :MFCD00004006
  •  :186.21 g/mol
  •  :Ambient
  • Pack type:
    Bottle