Chromomycin A3
Bottle
Catalog # APOSBIC1009-250MG
New Product
Supplier: Apollo Scientific
Chromomycin A3
Catalog # APOSBIC1009-250MG
Supplier: Apollo Scientific
New Product
CAS Number: 7059-24-7
Jump to:
Product Details & Documents
Antibiotic that can serve as a fluorescent DNA stain.
- CAS Number:7059-24-7
- Formula:C₅₇H₈₂O₂₆
- MDL Number:MFCD00043151
- Molecular Weight:1183.30 g/mol
- Melting Pt:182 °C
- Storage Temperature:Congelamento
- EINECS:230-348-0
- UN:3462
- ADR:6.1,I
Specifications
Specifications
- Tipo di conf.:Bottle
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



