Chromomycin A3
Bottle
Catalog # APOSBIC1009-50MG
New Product
Supplier: Apollo Scientific
Chromomycin A3
Catalog # APOSBIC1009-50MG
Supplier: Apollo Scientific
New Product
CAS Number: 7059-24-7
Jump to:
Product Details & Documents
Antibiotic that can serve as a fluorescent DNA stain.
- CAS Number:7059-24-7
- Formula:C₅₇H₈₂O₂₆
- MDL Number:MFCD00043151
- Molecular Weight:1183.30 g/mol
- Melting Pt:182 °C
- Storage Temperature:Freezer
- EINECS:230-348-0
- UN:3462
- ADR:6.1,I
Specifications
Specifications
- Pack type:Bottle



