Order Entry
Italy
Orders LinkContactUsLinkComponent
2-Chlorothioxanthone 98%
Bottle
  APOSOR471609-25G
New Product
2-Chlorothioxanthone 98%
  APOSOR471609-25G
New Product
 :  86-39-5
2-Chlorothioxanthone 98%
Bottle
 :  86-39-5
24,60 €
1 * 25 g
 :  2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
  •  :86-39-5
  •  :C₁₃H₇ClOS
  •  :MFCD00005067
  •  :246.72 g/mol
  •  :Ambient
  • Pack type:
    Bottle