Order Entry
Italy
Orders LinkContactUsLinkComponent
 

2-Chlorothioxanthone 98%

Bottle
Catalog # APOSOR471609-5G
New Product
Supplier:   Apollo Scientific
2-Chlorothioxanthone 98%
Catalog # APOSOR471609-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  86-39-5
2-Chlorothioxanthone 98%

Bottle

CAS Number:  86-39-5
Price
21,80 €
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  2-Chlorothioxanthen-9-one, 2-Chloro-9H-thioxanthen-9-one
  • CAS Number:86-39-5
  • Formula:C₁₃H₇ClOS
  • MDL Number:MFCD00005067
  • Molecular Weight:246.72 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications