Order Entry
Italy
Orders LinkContactUsLinkComponent
 

Quercetin ≥95%

Bottle
Catalog # APOSBUP18495-1G
Toxic
New Product
Supplier:   Apollo Scientific
Quercetin ≥95%
Catalog # APOSBUP18495-1G
Supplier:  Apollo Scientific
New Product
CAS Number:  117-39-5
Quercetin ≥95%

Bottle

CAS Number:  117-39-5
Price
63,70 €
Log in for availability
1 * 1 g
Product Details & Documents

Usage: Biochemical/ life science applications.

  • Store at –20 °C
Synonyms:  3-5-7-3-4-Pentahydroxyflavone, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-chromen-4-one, 2-(3,4-Dihydroxyphenyl)-3,5,7-trihydroxy-4H-1-benzopyran-4-one, 3,5,7,3',4'-Pentahydroxyflavone
  • CAS Number:117-39-5
  • Formula:C₁₅H₁₀O₇
  • MDL Number:MFCD00006828
  • Molecular Weight:302,24 g/mol
  • Storage Temperature:Freezer
  • EINECS:204-187-1
  • UN:2811
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.