Order Entry
Italy
Orders LinkContactUsLinkComponent
 

3-Mercaptopropionic acid 99%

Bottle
Catalog # APOSOR52782-2.5KG
New Product
Supplier:   Apollo Scientific
3-Mercaptopropionic acid 99%
Catalog # APOSOR52782-2.5KG
Supplier:  Apollo Scientific
New Product
CAS Number:  107-96-0
3-Mercaptopropionic acid 99%

Bottle

CAS Number:  107-96-0
Price
254,00 €
Log in for availability
1 * 2,5 kg
Product Details & Documents
  • CAS Number:107-96-0
  • Formula:C₃H₆O₂S
  • MDL Number:MFCD00004897
  • Molecular Weight:106.15 g/mol
  • Boiling Pt:110…111 °C/15mmHg
  • Melting Pt:17…19 °C
  • Density:1.218 g/cm³ (25 °C)
  • Flash Pt:124 °C
  • Storage Temperature:Ambient
  • UN:2922
  • ADR:8+6.1,II
Specifications

Specifications

  • Pack type:
    Bottle

Specifications