Order Entry
Italy
Orders LinkContactUsLinkComponent
3-Mercaptopropionic acid 99%

3-Mercaptopropionic acid 99%

Bottle
Catalog # APOSOR52782-100G
New Product
Supplier:   Apollo Scientific
3-Mercaptopropionic acid 99%
Catalog # APOSOR52782-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  107-96-0
3-Mercaptopropionic acid 99%

Bottle

CAS Number:  107-96-0
Price
24,60 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:107-96-0
  • Formula:C₃H₆O₂S
  • MDL Number:MFCD00004897
  • Molecular Weight:106.15 g/mol
  • Boiling Pt:110…111 °C/15mmHg
  • Melting Pt:17…19 °C
  • Density:1.218 g/cm³ (25 °C)
  • Flash Pt:124 °C
  • Storage Temperature:Ambient
  • UN:2922
  • ADR:8+6.1,II
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.