Order Entry
Italy
Orders LinkContactUsLinkComponent
3-Mercaptopropionic acid 99%

3-Mercaptopropionic acid 99%

Bottle
Codice catalogo APOSOR52782-25G
New Product
Nome del fornitore:   Apollo Scientific
3-Mercaptopropionic acid 99%
Codice catalogo APOSOR52782-25G
Nome del fornitore:  Apollo Scientific
New Product
Numero CAS:  107-96-0
3-Mercaptopropionic acid 99%

Bottle

Numero CAS:  107-96-0
Price
21,80 €
Log in for availability
1 * 25 g
Product Details & Documents
  • CAS Number:107-96-0
  • Formula:C₃H₆O₂S
  • MDL Number:MFCD00004897
  • Molecular Weight:106.15 g/mol
  • Boiling Pt:110…111 °C/15mmHg
  • Melting Pt:17…19 °C
  • Density:1.218 g/cm³ (25 °C)
  • Flash Pt:124 °C
  • Storage Temperature:Ambient
  • UN:2922
  • ADR:8+6.1,II
Specifications

Specifications

  • Pack type:
    Bottle

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.